Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: SPM558, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442301MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442301

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Primary or Secondary Primary
Immunogen Recombinant human Ig kappa chain
Quantity 0.1mg
Concentration 1mg/mL
Gene ID (Entrez) 3514
Monoclonal or Polyclonal Monoclonal
Regulatory Status RUO
Reconstitution Protein A purified
Clone SPM558
Cross Reactivity Human
Host Species Mouse
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Format Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity This MAb is specific to kappa light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30.
Purification Method Purified
Isotype IgG1, κ
Antigen Kappa Light Chain