Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: SPM558, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23442302MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23442302

  • / Each
 

Description & Specifications

Specifications

Antigen Kappa Light Chain
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Clone SPM558
Concentration 1mg/mL
Conjugate Unlabeled
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Murine
Immunogen Recombinant human Ig kappa chain
Isotype IgG1, κ
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3514
Test Specificity This MAb is specific to kappa light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. In mammals, the two light chains in an antibody are always identical, with only one type of light chain, kappa or lambda. The ratio of Kappa to Lambda is 70:30.
Reconstitution Protein A purified
Target Species Human