Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: SPM508, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23436605ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23436605

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Kappa Light Chain
Cross Reactivity Human
Format Purified
Immunogen Human B-Lymphoma Cells
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3514
Clone SPM508
Concentration 0.2mg/mL
Conjugate Unlabeled
Formulation PBS with 0.05% BSA.
Host Species Mouse
Quantity 0.5mL
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Test Specificity This MAb is specific to kappa light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. Does not react with Rat.
Reconstitution Protein A purified