missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: SPM508, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Kappa Light Chain
Clone SPM508
Concentration 1mg/mL
Immunogen Human B-Lymphoma Cells
Gene ID (Entrez) 3514
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23439601 Novus Biologicals Canada
NBP23439601MG
0.1mg Each for $384.00
NBP23439602 Novus Biologicals Canada
NBP23439602MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Kappa Light Chain
1mg/mL
3514
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1, κ
Purified
ELISA,Flow Cytometry,Immunohistochemistry (Frozen),Immunohistochemistry (Paraffin),Immunoprecipitation,Western Blot
SPM508
Human B-Lymphoma Cells
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified