Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: SPM508, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23439601MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23439601

  • / Each
 

Description & Specifications

Specifications

Antigen Kappa Light Chain
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Human B-Lymphoma Cells
Isotype IgG1, κ
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity This MAb is specific to kappa light chain of immunoglobulin and shows no cross-reaction with lambda light chain or any of the five heavy chains. Does not react with Rat.
Clone SPM508
Cross Reactivity Human
Purification Method Purified
Regulatory Status RUO
Gene ID (Entrez) 3514
Reconstitution Protein A purified
Concentration 1mg/mL
Conjugate Unlabeled