Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: KLC709, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23425901ML

 View all options for this product


For Research Use Only

Catalog No. NBP23425901


Description & Specifications


Antigen Kappa Light Chain
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Clone KLC709
Concentration 0.2mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P01601
Host Species Mouse
Immunogen Recombinant human Ig kappa chain
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3514
Reconstitution Protein A purified