Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: KLC709, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23466001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23466001

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Quantity 0.1mg
Antigen Kappa Light Chain
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P01601
Immunogen Recombinant human Ig kappa chain
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3514
Reconstitution Protein A purified
Clone KLC709
Concentration 1mg/mL
Conjugate Unlabeled
Host Species Mouse
Cross Reactivity Human