Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: KLC264, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23425501ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23425501

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Recombinant human Ig kappa chain
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Human
Host Species Mouse
Formulation PBS with 0.05% BSA.
Purification Method Purified
Regulatory Status RUO
Gene Accession No. P01601
Gene ID (Entrez) 3514
Clone KLC264
Concentration 0.2mg/mL
Antigen Kappa Light Chain
Monoclonal or Polyclonal Monoclonal
Format Purified
Quantity 0.1mL
Primary or Secondary Primary
Isotype IgG1, κ
Reconstitution Protein A purified
Conjugate Unlabeled