Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: KLC264, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23425505ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23425505

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Quantity 0.5mL
Conjugate Unlabeled
Format Purified
Formulation PBS with 0.05% BSA.
Antigen Kappa Light Chain
Monoclonal or Polyclonal Monoclonal
Isotype IgG1, κ
Regulatory Status RUO
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Purification Method Purified
Immunogen Recombinant human Ig kappa chain
Concentration 0.2mg/mL
Host Species Mouse
Clone KLC264
Primary or Secondary Primary
Reconstitution Protein A purified
Gene ID (Entrez) 3514
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P01601