Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: KLC264, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Kappa Light Chain
Clone KLC264
Concentration 1mg/mL
Immunogen Recombinant human Ig kappa chain
Gene Accession No. P01601
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23465901

 
novus biologicals canada
NBP23465901MG
0.1mg Each for $384.00

NBP23465902

 
novus biologicals canada
NBP23465902MG
0.2mg Each for $527.00
Description & Specifications

Specifications

Antigen Kappa Light Chain
Clone KLC264
Concentration 1mg/mL
Immunogen Recombinant human Ig kappa chain
Gene Accession No. P01601
Gene ID (Entrez) 3514
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot