Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: KLC264, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23465901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23465901

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Cross Reactivity Human
Format Purified
Gene Accession No. P01601
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3514
Concentration 1mg/mL
Antigen Kappa Light Chain
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Recombinant human Ig kappa chain
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mg
Reconstitution Protein A purified
Clone KLC264
Conjugate Unlabeled