Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: HP6053, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23425701ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23425701

  • / Each
 

Description & Specifications

Specifications

Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Antigen Kappa Light Chain
Formulation PBS with 0.05% BSA.
Gene Accession No. P01601
Host Species Mouse
Immunogen Purified human Ig kappa chain
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3514
Reconstitution Protein A purified
Clone HP6053
Concentration 0.2mg/mL
Conjugate Unlabeled
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.