Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: HP6053, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23425705ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23425705

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Host Species Mouse
Quantity 0.5mL
Antigen Kappa Light Chain
Immunogen Purified human Ig kappa chain
Concentration 0.2mg/mL
Cross Reactivity Human
Regulatory Status RUO
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Format Purified
Clone HP6053
Gene Accession No. P01601
Conjugate Unlabeled
Purification Method Purified
Primary or Secondary Primary
Isotype IgG1, κ
Reconstitution Protein A purified
Gene ID (Entrez) 3514