Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: HP6053 + L1C1, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Kappa Light Chain
Clone HP6053 + L1C1
Concentration 0.2mg/mL
Immunogen Purified human Ig kappa chain (HP6053) & Human B-Lymphoma Cells (L1C1)
Gene Accession No. P01601
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23425801

 
novus biologicals canada
NBP23425801ML
0.1mL Each for $139.00

NBP23425805

 
novus biologicals canada
NBP23425805ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Kappa Light Chain
Clone HP6053 + L1C1
Concentration 0.2mg/mL
Immunogen Purified human Ig kappa chain (HP6053) & Human B-Lymphoma Cells (L1C1)
Gene Accession No. P01601
Gene ID (Entrez) 3514
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting