Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: HP6053 + L1C1, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23425801ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23425801

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Host Species Mouse
Immunogen Purified human Ig kappa chain (HP6053) & Human B-Lymphoma Cells (L1C1)
Quantity 0.1mL
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3514
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Primary or Secondary Primary
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene Accession No. P01601
Reconstitution Protein A purified
Concentration 0.2mg/mL
Regulatory Status RUO
Clone HP6053 + L1C1
Antigen Kappa Light Chain