Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: HP6053 + L1C1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23464002MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23464002

  • / Each
 

Description & Specifications

Specifications

Antigen Kappa Light Chain
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified human Ig kappa chain (HP6053) & Human B-Lymphoma Cells (L1C1)
Isotype IgG1
Purification Method Purified
Quantity 0.2mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 3514
Clone HP6053 + L1C1
Concentration 1mg/mL
Conjugate Unlabeled
Gene Accession No. P01601
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Cross Reactivity Human