Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: HP6053 + L1C1, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23464001MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23464001

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Antigen Kappa Light Chain
Format Purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Isotype IgG1
Quantity 0.1mg
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Gene ID (Entrez) 3514
Conjugate Unlabeled
Cross Reactivity Human
Gene Accession No. P01601
Host Species Mouse
Immunogen Purified human Ig kappa chain (HP6053) & Human B-Lymphoma Cells (L1C1)
Purification Method Purified
Regulatory Status RUO
Reconstitution Protein A purified
Clone HP6053 + L1C1
Concentration 1mg/mL