Please login to your online account to display your discounted pricing

Mouse anti-Kappa Light Chain, Clone: HP6053, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23463902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23463902

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Monoclonal or Polyclonal Monoclonal
Immunogen Purified human Ig kappa chain
Gene Accession No. P01601
Gene ID (Entrez) 3514
Purification Method Purified
Format Purified
Antigen Kappa Light Chain
Primary or Secondary Primary
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Host Species Mouse
Reconstitution Protein A purified
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Clone HP6053
Quantity 0.2mg
Concentration 1mg/mL
Isotype IgG1, κ
Cross Reactivity Human