Please login to your online account to display your discounted pricing

Mouse anti-Kallikrein 3/PSA, Clone: SPM352, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23299301ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23299301

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Kallikrein 3/PSA
Gene Accession No. P07288
Host Species Mouse
Immunogen PSA from human sperm plasma
Regulatory Status RUO
Monoclonal or Polyclonal Monoclonal
Clone SPM352
Conjugate Unlabeled
Format Purified
Isotype IgG1, κ
Purification Method Purified
Quantity 0.1mL
Primary or Secondary Primary
Gene ID (Entrez) 354
Reconstitution Protein A purified
Concentration 0.2mg/mL
Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.