Please login to your online account to display your discounted pricing

Mouse anti-Kallikrein 3/PSA, Clone: SPM352, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23476802MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23476802

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Cross Reactivity Human
Conjugate Unlabeled
Isotype IgG1, κ
Format Purified
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Recognizes a single protein of 33-34kDa, identified as the prostate specific antigen (PSA). This MAb is highly specific to PSA and stains prostatic secretory and ductal epithelium in both normal and neoplastic tissues. PSA is a chymotrypsin-like serine protease (kallikrein family) exclusively produced by the prostate epithelium, and abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia.
Purification Method Purified
Regulatory Status RUO
Quantity 0.2mg
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Concentration 1mg/mL
Monoclonal or Polyclonal Monoclonal
Reconstitution Protein A purified
Immunogen PSA from human sperm plasma
Gene ID (Entrez) 354
Antigen Kallikrein 3/PSA
Clone SPM352
Gene Accession No. P07288