Please login to your online account to display your discounted pricing

Mouse anti-Kallikrein 3/PSA, Clone: SPM352, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23476801MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23476801

  • / Each
 

Description & Specifications

Specifications

Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Quantity 0.1mg
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Antigen Kallikrein 3/PSA
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P07288
Immunogen PSA from human sperm plasma
Purification Method Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 354
Test Specificity Recognizes a single protein of 33-34kDa, identified as the prostate specific antigen (PSA). This MAb is highly specific to PSA and stains prostatic secretory and ductal epithelium in both normal and neoplastic tissues. PSA is a chymotrypsin-like serine protease (kallikrein family) exclusively produced by the prostate epithelium, and abundant in seminal fluid. PSA can be detected in the sera of patients with prostatic carcinoma. It is predominantly complexed to a liver-derived serine protease inhibitor, alpha-1-antichymotrypsin (ACT). A higher proportion of serum PSA is complexed to ACT in prostate cancer than in benign prostate hyperplasia.
Reconstitution Protein A purified
Clone SPM352