Please login to your online account to display your discounted pricing

Mouse anti-Kallikrein 3/PSA, Clone: A67-B/E3, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23454902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23454902

  • / Each
 

Description & Specifications

Specifications

Antigen Kallikrein 3/PSA
Format Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.2mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P07288
Host Species Mouse
Immunogen PSA from human sperm plasma
Isotype IgG1, κ
Purification Method Purified
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 354
Reconstitution Protein A purified
Clone A67-B/E3
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human