Please login to your online account to display your discounted pricing

Mouse anti-Kallikrein 3/PSA, Clone: A67-B/E3, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23454901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23454901

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mg
Primary or Secondary Primary
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P07288
Conjugate Unlabeled
Format Purified
Concentration 1mg/mL
Isotype IgG1, κ
Antigen Kallikrein 3/PSA
Immunogen PSA from human sperm plasma
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Reconstitution Protein A purified
Gene ID (Entrez) 354
Clone A67-B/E3
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Regulatory Status RUO