missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-IRAK4, Clone: 6F8, Novus Biologicals™

$309.05 - $363.90

Specifications

Antigen IRAK4
Clone 6F8
Immunogen recombinant IRAK-4 transfected 293T cells
Gene ID (Entrez) 51135
Formulation Antibody in Tris-NaCl pH 8
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
DDX0340P05 Novus Biologicals Canada
DDX0340P100005MG
0.05mg Each for $309.05
DDX0340Y05 Novus Biologicals Canada
DDX0340A647005MG
0.05mg Each for $363.90
Specifications Description & Specifications

Specifications

IRAK4
recombinant IRAK-4 transfected 293T cells
Antibody in Tris-NaCl pH 8
Primary
Murine
Unlabeled
Ion Exchange Chromatography
6F8
51135
Monoclonal
Store at -20°C in the dark. Avoid freeze-thaw cycles.
IgG1
Purified
Flow Cytometry