Please login to your online account to display your discounted pricing

Mouse anti-IRAK4, Clone: 6F8, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0340A647005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0340Y05

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Host Species Mouse
Antigen IRAK4
Format Purified
Formulation Antibody in Tris-NaCl pH 8
Isotype IgG1
Quantity 0.05mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Immunogen recombinant IRAK-4 transfected 293T cells
Purification Method Ion Exchange Chromatography
Storage Requirements Store at -20°C in the dark. Avoid freeze-thaw cycles.
Gene ID (Entrez) 51135
Clone 6F8