Please login to your online account to display your discounted pricing

Mouse anti-IRAK4, Clone: 6F8, Novus Biologicals™


Manufacturer: novus biologicals canada  DDX0340P100005MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. DDX0340P05

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Quantity 0.05mg
Primary or Secondary Primary
Antigen IRAK4
Storage Requirements Store at -20°C. Avoid freeze-thaw cycles.
Conjugate Unlabeled
Immunogen IRAK-4 was used as immunogen.
Clone 6F8
Format Purified
Concentration 0.5mg/mL
Formulation Tris-NaCl pH 8
Cross Reactivity Canine
Cross Reactivity Human
Gene ID (Entrez) 51135
Regulatory Status RUO
Purification Method Ion Exchange Chromatography
Host Species Mouse
Monoclonal or Polyclonal Monoclonal
Gene Accession No. NP_001107654
Isotype IgG1