Please login to your online account to display your discounted pricing

Mouse anti-IPO-38, Clone: SPM260, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen IPO-38
Clone SPM260
Immunogen Spleen cells of a patient with hairy cell leukemia
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23445101

 
novus biologicals canada
NBP23445101ML
0.1mL Each for $139.00

NBP23445105

 
novus biologicals canada
NBP23445105ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen IPO-38
Clone SPM260
Immunogen Spleen cells of a patient with hairy cell leukemia
Formulation Culture Medium. Bioreactor concentrate.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Isotype IgM, κ
Conjugate Unlabeled
Format Concentrated
Purification Method Concentrated
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)