Please login to your online account to display your discounted pricing

Mouse anti-IPO-38, Clone: SPM260, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23445105ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23445105

  • / Each
 

Description & Specifications

Specifications

Antigen IPO-38
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Spleen cells of a patient with hairy cell leukemia
Isotype IgM, κ
Quantity 0.5mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Host Species Mouse
Purification Method Concentrated
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity Recognizes a protein of 14-16kDa, which is a novel nuclear antigen of proliferating cells.
Clone SPM260
Conjugate Unlabeled