Please login to your online account to display your discounted pricing

Mouse anti-IPO-38, Clone: SPM260, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23445101ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23445101

  • / Each
 

Description & Specifications

Specifications

Antigen IPO-38
Format Concentrated
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Spleen cells of a patient with hairy cell leukemia
Isotype IgM, κ
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Recognizes a protein of 14-16kDa, which is a novel nuclear antigen of proliferating cells.
Clone SPM260
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Formulation Culture Medium. Bioreactor concentrate.
Purification Method Concentrated