Please login to your online account to display your discounted pricing

Mouse anti-IPO-38, Clone: IPO-38, Novus Biologicals™

Manufacturer: novus biologicals canada  NBP23446901ML

 View all options for this product


For Research Use Only

Catalog No. NBP23446901


Description & Specifications


Antigen IPO-38
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Clone IPO-38
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Host Species Mouse
Immunogen Spleen cells of a patient with hairy cell leukemia
Isotype IgM, κ
Purification Method Concentrated
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal