Please login to your online account to display your discounted pricing

Mouse anti-IPO-38, Clone: IPO-38, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23446905ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23446905

  • / Each
 

Description & Specifications

Specifications

Antigen IPO-38
Format Concentrated
Formulation Culture Medium. Bioreactor concentrate.
Host Species Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Immunogen Spleen cells of a patient with hairy cell leukemia
Isotype IgM, κ
Purification Method Concentrated
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Clone IPO-38
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat