Please login to your online account to display your discounted pricing

Mouse anti-Involucrin, Clone: SY5, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Involucrin
Clone SY5
Concentration 0.2mg/mL
Immunogen Purified involucrin from human keratinocytes
Gene Accession No. P07476
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23426401

 
novus biologicals canada
NBP23426401ML
0.1mL Each for $139.00

NBP23426405

 
novus biologicals canada
NBP23426405ML
0.5mL Each for $321.00
Description & Specifications

Specifications

Antigen Involucrin
Clone SY5
Concentration 0.2mg/mL
Immunogen Purified involucrin from human keratinocytes
Gene Accession No. P07476
Gene ID (Entrez) 3713
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Canine
Cross Reactivity Gorilla
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Porcine
Cross Reactivity Primate
Isotype IgG1, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting