Please login to your online account to display your discounted pricing

Mouse anti-Involucrin, Clone: SY5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23426405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23426405

  • / Each
 

Description & Specifications

Specifications

Format Purified
Formulation PBS with 0.05% BSA.
Cross Reactivity Canine
Cross Reactivity Gorilla
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Porcine
Cross Reactivity Primate
Antigen Involucrin
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified involucrin from human keratinocytes
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3713
Test Specificity Mouse (Negative)
Reconstitution Protein A purified
Clone SY5
Concentration 0.2mg/mL
Conjugate Unlabeled
Gene Accession No. P07476