Please login to your online account to display your discounted pricing

Mouse anti-Involucrin, Clone: SY5, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23426401ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23426401

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Immunogen Purified involucrin from human keratinocytes
Isotype IgG1, κ
Gene ID (Entrez) 3713
Conjugate Unlabeled
Antigen Involucrin
Format Purified
Formulation PBS with 0.05% BSA.
Gene Accession No. P07476
Host Species Mouse
Purification Method Purified
Quantity 0.1mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Test Specificity Mouse (Negative)
Reconstitution Protein A purified
Clone SY5
Concentration 0.2mg/mL
Cross Reactivity Canine
Cross Reactivity Gorilla
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Porcine
Cross Reactivity Primate