Please login to your online account to display your discounted pricing

Mouse anti-Involucrin, Clone: SY5, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471901MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471901

  • / Each
 

Description & Specifications

Specifications

Antigen Involucrin
Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Cross Reactivity Canine
Cross Reactivity Gorilla
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Porcine
Cross Reactivity Primate
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Immunogen Purified involucrin from human keratinocytes
Isotype IgG1, κ
Quantity 0.1mg
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3713
Clone SY5
Gene Accession No. P07476
Purification Method Purified
Regulatory Status RUO
Test Specificity Mouse (Negative)
Concentration 1mg/mL
Conjugate Unlabeled
Reconstitution Protein A purified