Please login to your online account to display your discounted pricing

Mouse anti-Involucrin, Clone: SY5, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23471902MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23471902

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Canine
Cross Reactivity Gorilla
Cross Reactivity Human
Cross Reactivity Monkey
Cross Reactivity Porcine
Cross Reactivity Primate
Isotype IgG1, κ
Antigen Involucrin
Conjugate Unlabeled
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Reconstitution Protein A purified
Host Species Mouse
Immunogen Purified involucrin from human keratinocytes
Gene ID (Entrez) 3713
Primary or Secondary Primary
Quantity 0.2mg
Gene Accession No. P07476
Concentration 1mg/mL
Monoclonal or Polyclonal Monoclonal
Format Purified
Test Specificity Mouse (Negative)
Regulatory Status RUO
Purification Method Purified
Clone SY5
Formulation 10mM PBS. Does not contain BSA. Azide Free.