Please login to your online account to display your discounted pricing

Mouse anti-Involucrin, Clone: SPM259, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23435405ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23435405

  • / Each
 

Description & Specifications

Specifications

Formulation PBS with 0.05% BSA.
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Primary or Secondary Primary
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Primate
Antigen Involucrin
Format Purified
Immunogen Purified involucrin from human keratinocytes
Isotype IgG1, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3713
Test Specificity It recognizes a protein of 66kDa-170kDa, identified as involucrin. In Western blotting of cultured human keratinocytes, this MAb reacts with a 120kDa protein. It stains the involucrin in a variety of sizes: 170kDa in MCF-7 cells, a doublet of ∽115kDa and 150kDa in gorilla and owl monkey, 66kDa in dog, and a doublet of 105kDa in pig. Its epitope maps between codon 421-568 of human involucrin. Does not react with Mouse.
Reconstitution Protein A purified
Clone SPM259
Concentration 0.2mg/mL
Conjugate Unlabeled
Gene Accession No. P07476