Please login to your online account to display your discounted pricing

Mouse anti-Involucrin, Clone: SPM259, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440201MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440201

  • / Each
 

Description & Specifications

Specifications

Antigen Involucrin
Format Purified
Host Species Mouse
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Isotype IgG1, κ
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3713
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Primate
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Gene Accession No. P07476
Immunogen Purified involucrin from human keratinocytes
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Test Specificity It recognizes a protein of 66kDa-170kDa, identified as involucrin. In Western blotting of cultured human keratinocytes, this MAb reacts with a 120kDa protein. It stains the involucrin in a variety of sizes: 170kDa in MCF-7 cells, a doublet of ∽115kDa and 150kDa in gorilla and owl monkey, 66kDa in dog, and a doublet of 105kDa in pig. Its epitope maps between codon 421-568 of human involucrin. Does not react with Mouse.
Reconstitution Protein A purified
Clone SPM259
Concentration 1mg/mL