Please login to your online account to display your discounted pricing

Mouse anti-Involucrin, Clone: SPM259, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23440202MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23440202

  • / Each
 

Description & Specifications

Specifications

Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Antigen Involucrin
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Host Species Mouse
Immunogen Purified involucrin from human keratinocytes
Isotype IgG1, κ
Quantity 0.2mg
Regulatory Status RUO
Concentration 1mg/mL
Cross Reactivity Canine
Cross Reactivity Human
Cross Reactivity Porcine
Cross Reactivity Primate
Format Purified
Gene Accession No. P07476
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Gene ID (Entrez) 3713
Test Specificity It recognizes a protein of 66kDa-170kDa, identified as involucrin. In Western blotting of cultured human keratinocytes, this MAb reacts with a 120kDa protein. It stains the involucrin in a variety of sizes: 170kDa in MCF-7 cells, a doublet of ∽115kDa and 150kDa in gorilla and owl monkey, 66kDa in dog, and a doublet of 105kDa in pig. Its epitope maps between codon 421-568 of human involucrin. Does not react with Mouse.
Reconstitution Protein A purified
Clone SPM259
Conjugate Unlabeled