missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Mouse anti-Integrin beta 4/CD104, Clone: UM-A9, Azide Free, Novus Biologicals™

$384.00 - $527.00

Specifications

Antigen Integrin beta 4/CD104
Clone UM-A9
Concentration 1mg/mL
Immunogen Human squamous cell carcinoma (UM-SCC1)
Gene Accession No. P16144
View More Specs

Disclaimers:

For Research Use Only



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
NBP23450701 Novus Biologicals Canada
NBP23450701MG
0.1mg Each for $384.00
NBP23450702 Novus Biologicals Canada
NBP23450702MG
0.2mg Each for $527.00
Specifications Description & Specifications

Specifications

Integrin beta 4/CD104
1mg/mL
P16144
10mM PBS. Does not contain BSA. Azide Free.
Primary
Murine
Unlabeled
Purified
UM-A9
Human squamous cell carcinoma (UM-SCC1)
3691
Monoclonal
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a, κ
Purified
ELISA,Flow Cytometry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Frozen),Immunoprecipitation,Western Blot