Please login to your online account to display your discounted pricing

Mouse anti-Integrin beta 4/CD104, Clone: UM-A9, Azide Free, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23450701MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23450701

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Format Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Western Blotting
Gene ID (Entrez) 3691
Formulation 10mM PBS. Does not contain BSA. Azide Free.
Concentration 1mg/mL
Immunogen Human squamous cell carcinoma (UM-SCC1)
Antigen Integrin beta 4/CD104
Host Species Mouse
Regulatory Status RUO
Conjugate Unlabeled
Isotype IgG2a, κ
Purification Method Purified
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Clone UM-A9
Gene Accession No. P16144
Reconstitution Protein A purified
Quantity 0.1mg