Please login to your online account to display your discounted pricing

Mouse anti-Integrin beta 3/CD61, Clone: VI-PL2, Novus Biologicals™

$230.00 - $285.00

Specifications

Antigen Integrin beta 3/CD61
Clone VI-PL2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD61.
Gene Accession No. P05106
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP14339401

 
novus biologicals canada
NBP14339401MG
0.1mg Each for $285.00

NBP14339403

 
novus biologicals canada
NBP1433940025MG
0.025mg Each for $230.00
Description & Specifications

Specifications

Antigen Integrin beta 3/CD61
Clone VI-PL2
Concentration 0.5mg/mL
Immunogen The immunogen for this antibody was CD61.
Gene Accession No. P05106
Gene ID (Entrez) 3690
Formulation PBS [pH 7.2]
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate Unlabeled
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blot