Please login to your online account to display your discounted pricing

Mouse anti-Integrin beta 3/CD61, Clone: VI-PL2, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP1433940025MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP14339403

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Human
Immunogen The immunogen for this antibody was CD61.
Regulatory Status RUO
Antigen Integrin beta 3/CD61
Formulation PBS [pH 7.2]
Clone VI-PL2
Gene Accession No. P05106
Isotype IgG1, κ
Conjugate Unlabeled
Quantity 0.025mg
Concentration 0.5mg/mL
Monoclonal or Polyclonal Monoclonal
Host Species Mouse
Format Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Western Blotting
Test Specificity The VI-PL2 monoclonal antibody reacts with human CD61, also known as integrin beta 3 and GPIIIa. This VI-PL2 antibody has also been shown to crossreact with canine and several non-human primate species, including baboon, rhesus, and cynomologus monkey.
Storage Requirements Store at 4°C. Do not freeze.
Purification Method Affinity Purified
Primary or Secondary Primary
Gene ID (Entrez) 3690