Please login to your online account to display your discounted pricing

Mouse anti-Integrin beta 3/CD61, Clone: VI-PL2, FITC, Novus Biologicals™

$131.00 - $252.00

Specifications

Antigen Integrin beta 3/CD61
Clone VI-PL2
Immunogen The immunogen for this antibody was CD61.
Gene Accession No. P05106
Gene ID (Entrez) 3690
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP200247

 
novus biologicals canada
NBP200247100TESTS
100 Tests Each for $252.00

NBP20024725

 
novus biologicals canada
NBP20024725TESTS
25 Tests Each for $131.00
Description & Specifications

Specifications

Antigen Integrin beta 3/CD61
Clone VI-PL2
Immunogen The immunogen for this antibody was CD61.
Gene Accession No. P05106
Gene ID (Entrez) 3690
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Murine
Isotype IgG1, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry