Please login to your online account to display your discounted pricing

Mouse anti-Integrin beta 1/CD29, Clone: TS2/16, FITC, Novus Biologicals™

$128.00 - $128.00

Specifications

Antigen Integrin beta 1/CD29
Clone TS2/16
Immunogen The immunogen was the full length version of the protein: CD29 (TS2/16)
Gene Accession No. P05556
Gene ID (Entrez) 3688
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP19999825

 
novus biologicals canada
NBP19999825TESTS
25 Tests Each for $128.00
Description & Specifications

Specifications

Antigen Integrin beta 1/CD29
Clone TS2/16
Immunogen The immunogen was the full length version of the protein: CD29 (TS2/16)
Gene Accession No. P05556
Gene ID (Entrez) 3688
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C. Do not freeze.
Host Species Mouse
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Isotype IgG1, κ
Conjugate FITC
Format Affinity Purified
Purification Method Affinity Purified
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunoprecipitation