Please login to your online account to display your discounted pricing

Mouse anti-Integrin beta 1/CD29, Clone: TS2/16, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP19999825TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP19999825

  • / Each
 

Description & Specifications

Specifications

Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunoprecipitation
Conjugate FITC
Primary or Secondary Primary
Quantity 25 Tests
Isotype IgG1, κ
Antigen Integrin beta 1/CD29
Gene ID (Entrez) 3688
Monoclonal or Polyclonal Monoclonal
Gene Accession No. P05556
Test Specificity Canine reactivity reported in scientific literature (PMID: 7698991).
Clone TS2/16
Regulatory Status RUO
Purification Method Affinity Purified
Format Affinity Purified
Host Species Mouse
Immunogen The immunogen was the full length version of the protein: CD29 (TS2/16)
Storage Requirements Store at 4°C. Do not freeze.