Please login to your online account to display your discounted pricing

Mouse anti-Integrin beta 1/CD29, Clone: TS2/16, FITC, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP199998100TESTS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP199998

  • / Each
 

Description & Specifications

Specifications

Applications Flow Cytometry
Applications Functional Assay
Applications Immunohistochemistry
Applications Immunoprecipitation
Quantity 100 Tests
Primary or Secondary Primary
Cross Reactivity Bovine
Cross Reactivity Canine
Cross Reactivity Human
Antigen Integrin beta 1/CD29
Format Affinity Purified
Isotype IgG1, κ
Purification Method Affinity Purified
Regulatory Status RUO
Storage Requirements Store at 4°C. Do not freeze.
Gene ID (Entrez) 3688
Conjugate FITC
Gene Accession No. P05556
Host Species Mouse
Immunogen The immunogen was the full length version of the protein: CD29 (TS2/16)
Monoclonal or Polyclonal Monoclonal
Test Specificity Canine reactivity reported in scientific literature (PMID: 7698991).
Clone TS2/16