Please login to your online account to display your discounted pricing

Mouse anti-Integrin beta 1/CD29, Clone: P4C10, Novus Biologicals™

$139.00 - $329.00

Specifications

Antigen Integrin beta 1/CD29
Clone P4C10
Concentration 1.0mg/mL
Immunogen HT1080 cells human fibrosarcoma
Gene Accession No. P05556
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP236561

 
novus biologicals canada
NBP236561
0.1mL Each for $329.00

NBP236561SS

 
novus biologicals canada
NBP236561SS
0.025mL Each for $139.00
Description & Specifications

Specifications

Antigen Integrin beta 1/CD29
Clone P4C10
Concentration 1.0mg/mL
Immunogen HT1080 cells human fibrosarcoma
Gene Accession No. P05556
Gene ID (Entrez) 3688
Formulation PBS. Protein G purified.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Murine
Isotype IgG1
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications Blocking
Applications ELISA
Applications Flow Cytometry
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Applications Neutralization