Please login to your online account to display your discounted pricing

Mouse anti-Integrin alpha M/CD11b, Clone: C11b/660, Novus Biologicals™

$139.00 - $321.00

Specifications

Antigen Integrin alpha M/CD11b
Clone C11b/660
Concentration 0.2mg/mL
Immunogen Recombinant human CD11b protein
Gene Accession No. P11215
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

NBP23426205

 
novus biologicals canada
NBP23426205ML
0.5mL Each for $321.00

NBP23426201

 
novus biologicals canada
NBP23426201ML
0.1mL Each for $139.00
Description & Specifications

Specifications

Antigen Integrin alpha M/CD11b
Clone C11b/660
Concentration 0.2mg/mL
Immunogen Recombinant human CD11b protein
Gene Accession No. P11215
Gene ID (Entrez) 3684
Formulation PBS with 0.05% BSA.
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Host Species Mouse
Cross Reactivity Human
Isotype IgG2b, κ
Conjugate Unlabeled
Format Purified
Purification Method Purified
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation