Please login to your online account to display your discounted pricing

Mouse anti-Integrin alpha M/CD11b, Clone: C11b/660, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP23426205ML

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP23426205

  • / Each
 

Description & Specifications

Specifications

Antigen Integrin alpha M/CD11b
Format Purified
Host Species Mouse
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Conjugate Unlabeled
Cross Reactivity Human
Formulation PBS with 0.05% BSA.
Gene Accession No. P11215
Applications ELISA
Applications Flow Cytometry
Applications Functional Assay
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Frozen)
Applications Immunoprecipitation
Immunogen Recombinant human CD11b protein
Isotype IgG2b, κ
Purification Method Purified
Quantity 0.5mL
Regulatory Status RUO
Storage Requirements Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Gene ID (Entrez) 3684
Reconstitution Protein A purified
Clone C11b/660
Concentration 0.2mg/mL